o-Xylen for syntese, Sigma-Aldrich®

Leverandør: Merck
Danger


M-Clarity™ quality level = MQ200

8.08697.1000 8.08697.2500 8.08697.6010
8.08697.1000EA 1050 NOK
8.08697.1000 8.08697.2500 8.08697.6010
o-Xylen for syntese, Sigma-Aldrich®
o-Xylen
Formel: C₈H₁₀
Kokepunkt: 144 °C (1013 hPa)
Smeltepunkt: –25 °C
Tetthet: 0,88 g/cm³ (20 °C)
Flash punkt: 30 °C (closed cup)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00008519
CAS-nummer: 95-47-6
EINECS: 202-422-2
FN: 1307
ADR: 3,III
Merck Indeks: 12,10214
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5dd6c62d1c62b6e7f71df7e157aae77162b3bcf1&vid=3ec8c09bb94f634e9f90b6f9e8320cd626f276d0&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Solubility 0.18 g/l (20 °C)
Melting Point -25 °C
Molar Mass 106.17 g/mol
Boiling Point 144.4 °C (1013 hPa)
Vapor Pressure 7 hPa (20 °C)
Flash Point 30 °C
Explosion Limit 1.0 - 7.6 %(V)
Density 0.88 g/cm3 (20 °C)
Ignition Point 465 °C
Evaporation Number 13.5


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR