o-kresol for syntese, Sigma-Aldrich®

Leverandør: Merck
Danger


M-Clarity™ quality level = MQ200

8.09692.0100 8.09692.0500 8.09692.1000 8.09692.2500 8.09692.9025
8.09692.0100EA 368 NOK
8.09692.0100 8.09692.0500 8.09692.1000 8.09692.2500 8.09692.9025
o-kresol for syntese, Sigma-Aldrich®
o-kresol
Formel: C₇H₈O
Kokepunkt: 191 °C (1013 hPa)
Smeltepunkt: 30…34 °C
Tetthet: 1,047 g/cm³ (20 °C)
Flash punkt: 81 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00002226
CAS-nummer: 95-48-7
EINECS: 202-423-8
FN: 3455
ADR: 6.1 (8),II
Merck Indeks: 13,02604

Bestill nå

Spesifikasjon Testresultater

Solubility 20 g/l (20 °C)
Melting Point 31 °C
Boiling Point 191 - 192 °C (1013 hPa)
Vapor Pressure 0.24 hPa (20 °C)
Flash Point 81 °C
Refractive Index 1.553 (20 °C, 589 nm)
Explosion Limit 1.3 %(V)
Density 1.046 g/cm3 (20 °C)
pH 4.8 (20 g/l, H2O, 20 °C)
Ignition Point 555 °C
Synonyms 2-Hydroxytoluene, 2-Methylphenol
Hill Formula C7H8O


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR