Methyltrimethoxysilane for synthesis, Sigma-Aldrich®

Leverandør: Merck
Danger

Synonymer: Trimethoxy(methyl)silane

M-Clarity™ quality level = MQ200

8.14178.0250
8.14178.0250EA 694 NOK
8.14178.0250
Methyltrimethoxysilane for synthesis, Sigma-Aldrich®
Methyltrimethoxysilane
Formel: C₄H₁₂O₃Si
Kokepunkt: 102 °C (1013 hPa)
Smeltepunkt: 11 °C
Tetthet: 0,955 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00008342
CAS-nummer: 1185-55-3
EINECS: 214-685-0
FN: 1993
ADR: 3,II

Bestill nå

Spesifikasjon Testresultater

Solubility (20 °C) (decomposition)
Molar Mass 136.22 g/mol
Boiling Point 102 - 103 °C (1013 hPa)
Flash Point 9 °C
Refractive Index 1.3646 (20 °C, 589 nm)
Density 0.96 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR