Bis(2-ethylhexyl)phosphate zS

Leverandør: Merck
Danger

Synonymer: Bis(2-ethylhexyl) hydrogen phosphate, Di-(2-ethylhexyl)phosphoric acid

MQ200

8.14181.0250 8.14181.1000
8.14181.0250EA 405 NOK
8.14181.0250 8.14181.1000
Bis(2-ethylhexyl)phosphate zS
Bis(2-ethylhexyl)phosphate
Formel: C₁₆H₃₅O₄P
MW: 322.43 g/mol
MDL nummer: MFCD00009492
CAS-nummer: 298-07-7
EINECS: 206-056-4
FN: 1902
ADR: 8 III

Bestill nå

Spesifikasjon Testresultater

Solubility  - 1 g/l (20 °C)
Melting Point -50 °C
Molar Mass 322.42 g/mol
Boiling Point 48 °C (16 hPa)
Vapor Pressure  - 0.1 hPa (20 °C)
Flash Point 150 °C
Refractive Index 1.443 (20 °C, 589 nm)
Density 0.97 g/cm3 (20 °C)
pH 3 ( 1 g/l, H2O)
Ignition Point 300 °C
Synonyms HDEHP, Phosphoric acid bis (2-ethylhexyl) ester, Bis(2-ethylhexyl) phosphoric acid
Hill Formula C16H35O4P
Chemical Formula (CH3(CH2)3CH(C2H5)CH2O)2POOH

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR