Pyridinium-p-toluenesulphonate, Sigma-Aldrich®

Leverandør: Merck

Synonymer: Pyridin-1-ium 4-methylbenzenesulphonate

M-Clarity™ quality level = MQ200

8.14330.0025 8.14330.0100
8.14330.0025EA 739 NOK
8.14330.0025 8.14330.0100
Pyridinium-p-toluenesulphonate, Sigma-Aldrich®
Pyridinium-p-toluenesulphonate
Formel: C₁₂H₁₃NO₃S
MDL nummer: MFCD00013108
CAS-nummer: 24057-28-1
EINECS: 246-002-7

Bestill nå

Spesifikasjon Testresultater

Solubility (20 °C) soluble
Melting Point 112 - 119 °C (dry substance)
Molar Mass 251.30 g/mol
Hill Formula C12H13NO3S
Chemical Formula 4-(CH3)C6H4SO3H * C5H5N


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR