Hydroxylamine 50% vandig løsning for syntese, Sigma-Aldrich®

Leverandør: Merck
Danger


M-Clarity™ quality level = MQ200

8.14441.0100 8.14441.0500
8.14441.0100EA 362 NOK
8.14441.0100 8.14441.0500
Hydroxylamine 50% vandig løsning for syntese, Sigma-Aldrich®
Hydroxylamine
Formel: H₃NO
Tetthet: 1,12 g/cm3 (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00044522
CAS-nummer: 7803-49-8
EINECS: 232-259-2
FN: 3266
ADR: 8,III

Bestill nå

Spesifikasjon Testresultater

Assay (manganometric) 47.0 - 53.0 %
Density (d 20 °C/ 4 °C) 1.120 - 1.123
Vapor Pressure 14 hPa (20 °C)
Density 1.12 g/cm3 (20 °C)
pH 10.6 (H2O, 20 °C)
Ignition Point 215 °C
Viscosity kinematic 1.435 mm2/s (25 °C)
Hazard Class 9
Hazard Class Description Miscellaneous hazardous material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR