Bis(8-hydroxyquinolinium)sulphate monohydrate, Sigma-Aldrich®

Leverandør: Merck
Warning

Synonymer: Quinolin-8-ol sulphate hydrate, Quinolin-8-ol hemisulphate hemihydrate, 8-Hydroxyquinoline sulphate monohydrate, 8-Hydroxyquinolinium hemisulphate hemihydrate, 8-Hydroxyquinoline hemisulphate salt hemihydrate

M-Clarity™ quality level = MQ200

8.14528.0100
8.14528.0100EA 530 NOK
8.14528.0100
Bis(8-hydroxyquinolinium)sulphate monohydrate, Sigma-Aldrich®
Bis(8-hydroxyquinolinium)sulphate monohydrate
Formel: C₁₈H₁₆N₂O₆S·1H₂O
MDL nummer: MFCD00149610
CAS-nummer: 207386-91-2
EINECS: 205-137-1

Bestill nå

Spesifikasjon Testresultater

Solubility (20 °C) soluble
Melting Point 177 °C
Molar Mass 406.41 g/mol
pH 3.2 (H2O, 20 °C)

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR