Propylene glycol diacetate for synthesis, Sigma-Aldrich®

Leverandør: Merck

Synonymer: Propane-1,2-diyl diacetate, 1,2-Propylene glycol diacetate, 1,2-Propanediol diacetate

M-Clarity™ quality level = MQ200

8.14588.1000
8.14588.1000EA 617 NOK
8.14588.1000
Propylene glycol diacetate for synthesis, Sigma-Aldrich®
Propylene glycol diacetate
Formel: C₇H₁₂O₄
MDL nummer: MFCD00026201
CAS-nummer: 623-84-7
EINECS: 210-817-6

Bestill nå

Spesifikasjon Testresultater

Solubility 90 g/l (20 °C)
Molar Mass 160.16 g/mol
Boiling Point 190 - 191 °C (1013 hPa)
Vapor Pressure  - 1 hPa (20 °C)
Flash Point 87 °C
Explosion Limit 2.8 - 12.7 %(V)
Density 1.05 g/cm3 (20 °C)
Ignition Point 431 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR