Natriumperoksid for synthesis, Sigma-Aldrich®

Leverandør: Merck
Danger

Synonymer: di-Sodium peroxide

M-Clarity™ quality level = MQ200

8.14606.0100
8.14606.0100EA 717 NOK
8.14606.0100
Natriumperoksid for synthesis, Sigma-Aldrich®
Natriumperoksid
Formel: Na₂O₂
Smeltepunkt: 660 °C
Tetthet: 2,805 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003497
CAS-nummer: 1313-60-6
EINECS: 215-209-4
FN: 1504
ADR: 5.1,I
Merck Indeks: 13,08728

Bestill nå

Spesifikasjon Testresultater

Solubility 100 g/l (20 °C) (decomposition)
Melting Point 660 °C
Molar Mass 77.98 g/mol
Bulk Density 1138 kg/m3
Density 2.8 g/cm3 (20 °C)
pH 12.8 (100 g/l, H2O, 20 °C)
Hazard Class 5.1
Hazard Class Description Oxidizer


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR