Antimon(III)klorid 99,0 Iodometry zS, Sigma-Aldrich®

Leverandør: Merck
Danger

Synonymer: Antimony trichloride

MQ200

8.14656.9045
8.14656.9045EA 0 NOK
8.14656.9045
Antimon(III)klorid 99,0 Iodometry zS, Sigma-Aldrich®
Antimon(III)klorid
Formel: SbCl₃
MW: 228.12 g/mol
Kokepunkt: 223.5
Smeltepunkt: 73
MDL nummer: MFCD00011212
CAS-nummer: 10025-91-9
FN: 1733
ADR: 8 II
Merck Indeks: 13,00713

Bestill nå

Spesifikasjon Testresultater

Solubility 931 g/l (20 °C) Hydrolysis
Melting Point 73 °C
Molar Mass 228.1 g/mol
Bulk Density 1600 kg/m3
Boiling Point 233 °C (1013 hPa)
Vapor Pressure 0.16 hPa (20 °C)
Density 3.14 g/cm3 (20 °C)
pH (H2O) acidic
Synonyms Antimonous acid, Antimony trichloride
Hill Formula Cl3Sb
Chemical Formula SbCl3


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR