Tetrakis(triphenylphosphine)palladium(0) for synthesis, Sigma-Aldrich®

Leverandør: Merck
Warning

Synonymer: Pd(PPh3)4

M-Clarity™ quality level = MQ200

8.14761.0005 8.14761.0001 8.14761.0025 8.14761.8500
8.14761.0005EA 3190 NOK
8.14761.0005 8.14761.0001 8.14761.0025 8.14761.8500
Tetrakis(triphenylphosphine)palladium(0) for synthesis, Sigma-Aldrich®
Tetrakis(triphenylphosphine)palladium(0)
Formel: C₇₂H₆₀P₄Pd
Smeltepunkt: 103…107 °C
Tetthet: 1 g/cm³ (20 °C)
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00010012
CAS-nummer: 14221-01-3
EINECS: 238-086-9

Bestill nå

Spesifikasjon Testresultater

Solubility (20 °C) insoluble
Melting Point 103 - 107 °C
Molar Mass 1155.58 g/mol
Hill Formula C72H60P4Pd
Chemical Formula Pd(P(C6H5)3)4


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR