2,4,5-Triamino-6-hydroxypyrimidine sulphate for synthesis, Sigma-Aldrich®

Leverandør: Merck

Synonymer: 2,5,6-Triaminopyrimidin-4(1H)-one sulphate, 4-Hydroxy-2,5,6-triaminopyrimidine sulphate, 6-Hydroxy-2,4,5-triaminopyrimidine sulphate

M-Clarity™ quality level = MQ200

8.14764.0025
8.14764.0025EA 510 NOK
8.14764.0025
2,4,5-Triamino-6-hydroxypyrimidine sulphate for synthesis, Sigma-Aldrich®
2,4,5-Triamino-6-hydroxypyrimidine sulphate
Formel: C₄H₉N₅O₅S
MDL nummer: MFCD00012785
CAS-nummer: 39267-74-8
EINECS: 254-393-0

Bestill nå

Spesifikasjon Testresultater

Solubility 2 g/l (20 °C)
Melting Point 350 °C
Bulk Density 480 kg/m3
pH 2 - 3 (2 g/l, H2O, 20 °C)
Hill Formula C4H9N5O5S
Chemical Formula C4H7N5O . x H2SO4


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR