5-(4-Fluorophenyl)-3-pyridinecarbaldehyde zS

Leverandør: Merck

Synonymer: 5-(4-Fluorophenyl)nicotinaldehyde

8.14808.0010
8.14808.0010EA 0 NOK
8.14808.0010
5-(4-Fluorophenyl)-3-pyridinecarbaldehyde zS
5-(4-Fluorophenyl)-3-pyridinecarbaldehyde
Formel: C₁₂H₈FNO
MW: 201.2000 g/mol
MDL nummer: MFCD02093924
CAS-nummer: 381684-96-4

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR