Acetic acid-[D1]

Leverandør: Merck
Danger

Synonymer: Ethanoic acid-D1

8.15035.0050
8.15035.0050EA 0 NOK
8.15035.0050
Acetic acid-[D1]
Acetic acid-[D1]
Formel: C₂H₃DO₂
MW: 61.04 g/mol
Kokepunkt: 116
Smeltepunkt: 17
Tetthet: 1.05 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00051050
CAS-nummer: 758-12-3
EINECS: 212-059-1

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR