L(+)-Leucin for syntese, Sigma-Aldrich®

Leverandør: Merck

Synonymer: (S)-(+)-Leucine, Leu, H-Leu-OH, Leucine, L-Leucine

M-Clarity™ quality level = MQ200

8.16009.0100
8.16009.0100EA 655 NOK
8.16009.0100
L(+)-Leucin for syntese, Sigma-Aldrich®
L(+)-Leucin
Formel: C₆H₁₃NO₂
MW: 131,17 g/mol
Smeltepunkt: 300 °C
Tetthet: 1,165 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00002617
CAS-nummer: 61-90-5
EINECS: 200-522-0
Merck Indeks: 14,05451

Bestill nå

Spesifikasjon Testresultater

Solubility 24 g/l (20 °C)
Melting Point 300 °C (decomposition)
Molar Mass 131.18 g/mol
Bulk Density 300 - 400 kg/m3
Density 1.29 g/cm3 (20 °C)
pH 5.5 - 6.5 (20 g/l, H2O, 20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR