Triallyl isocyanurate zS, Sigma-Aldrich®

Leverandør: Merck
Warning

Synonymer: 1,3,5-Triallyl-1,3,5-triazine-2,4,6(1H,3H,5H)-trione, Triallyl-s-triazine-2,4,6(1H,3H,5H)-trione, Isocyanuric Acid Triallyl Ester, 1,3,5-Triallyl-1,3,5-triazinane-2,4,6-trione

MQ200

8.16113.0100
8.16113.0100EA 850 NOK
8.16113.0100
Triallyl isocyanurate zS, Sigma-Aldrich®
Triallyl isocyanurate
Formel: C₁₂H₁₅N₃O₃
MW: 249.27 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00006554
CAS-nummer: 1025-15-6
EINECS: 213-834-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ba173febf45c799ccb4dac91462ae7762eff24d2&vid=ba173febf45c799ccb4dac91462ae7762eff24d2&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Solubility 3.7 g/l (30 °C)
Melting Point 25 °C
Molar Mass 249.27 g/mol
Boiling Point 150 °C (5.3 hPa)
Vapor Pressure 3.5 hPa (143 °C)
Flash Point 160 °C
Density 1.16 g/cm3 (20 °C)
Viscosity Kinematic 78.2 mm2/s (30 °C)
Storage +15...+25 °C
Assay ≥98,0% (a/a)
Identity Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR