Trifenylfosfin selenid for syntese, Sigma-Aldrich®

Leverandør: Merck
Danger


M-Clarity™ quality level = MQ200

8.18054.0050
8.18054.0050EA 1200 NOK
8.18054.0050
Trifenylfosfin selenid for syntese, Sigma-Aldrich®
Trifenylfosfin selenid
Formel: C₁₈H₁₅PSe
MW: 341.25 g/mol
Smeltepunkt: 186…189 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00013477
CAS-nummer: 3878-44-2
EINECS: 223-406-1
FN: 3467
ADR: 6.1 II

Bestill nå

Spesifikasjon Testresultater

Solubility (20 °C) insoluble
Melting Point 186 - 189 °C
Molar Mass 341.25 g/mol
Hazard Class 6.1
Hazard Class Description Toxic materials

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR