(S)-(-)-β-Citronellol for syntese, Sigma-Aldrich®

Leverandør: Merck
Warning


M-Clarity™ quality level = MQ200

8.18378.0025 8.18378.0010 8.18378.9025 8.18378.8000
8.18378.0025EA 895 NOK
8.18378.0025 8.18378.0010 8.18378.9025 8.18378.8000
(S)-(-)-β-Citronellol for syntese, Sigma-Aldrich®
(S)-(-)-β-Citronellol
Formel: C₁₀H₂₀O
Tetthet: 0,855 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00063214
CAS-nummer: 7540-51-4
EINECS: 231-415-7
FN: 3082
ADR: 9,III

Bestill nå

Spesifikasjon Testresultater

Solubility (20 °C) practically insoluble
Molar Mass 156.27 g/mol
Boiling Point 109 - 111 °C (13 hPa)
Flash Point 101 °C DIN 51758
Refractive Index 1.4562 (20 °C, 589 nm)
Density 0.855 g/cm3 (20 °C)
Ignition Point 240 °C
Synonyms (S)-(-)-3,7-Dimethyl-6-octen-1-ol, ß-Rhodinol
Hill Formula C10H20O

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR