Tyramine hydrochloride, Sigma-Aldrich®

Leverandør: Merck

Synonymer: 2-(4-Hydroxyphenyl)ethylammonium chloride, 4-(2-Aminoethyl)phenol hydrochloride, 4-Hydroxyphenethylammonium chloride

M-Clarity™ quality level = MQ200

8.18444.0025
8.18444.0025EA 1080 NOK
8.18444.0025
Tyramine hydrochloride, Sigma-Aldrich®
Tyramine hydrochloride
Formel: C₈H₁₂ClNO
Smeltepunkt: 270…272 °C
MDL nummer: MFCD00012901
CAS-nummer: 60-19-5
EINECS: 200-462-5

Bestill nå

Spesifikasjon Testresultater

Solubility (20 °C) soluble
Melting Point 268 - 269 °C
Molar Mass 173.64 g/mol
Synonyms 4-(2-Aminoethyl)phenol hydrochloride, Tyramine hydrochloride
Hill Formula C8H12ClNO
Chemical Formula C8H11NO * HCl


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR