(+)-Dimethyl-D-malate for synthesis

Leverandør: Merck

Synonymer: D-(+)-Malic acid dimethyl ester, Dimethyl D-(+)-Malate, D-(+)-Apple acid dimethyl ester, (R)-Dimethyl 2-hydroxysuccinate

8.18523.0001 8.18523.0005
8.18523.0001EA 0 NOK
8.18523.0001 8.18523.0005
(+)-Dimethyl-D-malate for synthesis
(+)-Dimethyl-D-malate
Formel: C₆H₁₀O₅
MW: 162.1406 g/mol
MDL nummer: MFCD00066216
CAS-nummer: 70681-41-3

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR