2-Decanon for syntese, Sigma-Aldrich®

Leverandør: Merck

Synonymer: Decan-2-one

M-Clarity™ quality level = MQ200

8.18795.0025
8.18795.0025EA 1000 NOK
8.18795.0025
2-Decanon for syntese, Sigma-Aldrich®
2-Decanone
Formel: C₁₀H₂₀O
MW: 156,27 g/mol
Kokepunkt: 211 °C (1013 hPa)
Smeltepunkt: 3 °C
Tetthet: 0,82 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00009571
CAS-nummer: 693-54-9
EINECS: 211-752-6

Bestill nå

Spesifikasjon Testresultater

Solubility (20 °C) insoluble
Melting Point 3 °C
Molar Mass 156.26 g/mol
Boiling Point 209 - 212 °C
Flash Point 86 °C
Refractive Index 1.4253 (20 °C, 589 nm)
Density 0.82 g/cm3 (20 °C)
Synonyms Methyl octyl ketone
Hill Formula C10H20O
Chemical Formula CH3(CH2)7COCH3

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR