iso-Amylalkohol for synthesis, Sigma-Aldrich®

Leverandør: Merck
Warning

Synonymer: Isopentyl alcohol, iso-Pentanol, 3-Methyl-1-butanol, 3-Methylbutanol

M-Clarity™ quality level = MQ200

8.18969.1000
8.18969.1000EA 857 NOK
8.18969.1000
iso-Amylalkohol for synthesis, Sigma-Aldrich®
iso-Amylalkohol
Formel: (CH₃)₂CHCH₂CH₂OH
MW: 88.15 g/mol
Kokepunkt: 130
Smeltepunkt: –117 °C
Tetthet: 0.812 g/cm³ (20 °C)
Flash punkt: 42
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00002934
CAS-nummer: 123-51-3
EINECS: 204-633-5
FN: 1105
ADR: 3 III
Merck Indeks: 13,05214
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cd7b89cc1890b5d4b2083195c30c0d1047c32e9a&vid=170c60e26c7b4edab6e0686f91d04e676da16e6d&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Solubility 25 g/l (20 °C)
Melting Point -117 °C
Molar Mass 88.15 g/mol
Boiling Point 131 °C (1013 hPa)
Vapor Pressure 3 hPa (20 °C)
Flash Point 45 °C
Explosion Limit 1.2 - 8 %(V)
Density 0.81 g/cm3 (20 °C)
pH 7 (25 g/l, H2O, 20 °C)
Ignition Point 335 °C
Saturation concentration 11 g/m3 (20 °C) Air
Hazard Class 3
Hazard Class Description Flammable Liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR