1-Hydroxy-2-pyridinthione sodium salt 40% water

Leverandør: Merck
Warning


MQ200

8.19054.0250
8.19054.0250EA 1100 NOK
8.19054.0250
1-Hydroxy-2-pyridinthione sodium salt 40% water
1-Hydroxy-2-pyridinthione sodium salt
Formel: C₅H₄NNaOS
MW: 149.143 g/mol
Lagringstemperatur: Romtemperatur
CAS-nummer: 15922-78-8
FN: 3082
ADR: 9 III

Bestill nå

Spesifikasjon Testresultater

Solubility (20 °C) miscible in all proportions
Boiling Point 100 °C (1013 hPa)
Density 1.22 g/cm3 (20 °C)
pH 8.5 - 10.5 (H2O, 20 °C)
Storage temperature +15...+25 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR