Anisol, Sigma-Aldrich®

Leverandør: Merck
Danger


M-Clarity™ quality level = MQ200

8.19243.6190
8.19243.6190EA 0 NOK
8.19243.6190
Anisol, Sigma-Aldrich®
Anisol
Formel: C₇H₈O
Kokepunkt: 154 °C (1013 hPa)
Smeltepunkt: –37 °C
Tetthet: 0,9961 g/cm³ (20 °C)
Flash punkt: 44 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00008354
CAS-nummer: 100-66-3
EINECS: 202-876-1
FN: 2222
ADR: 3,III
Merck Indeks: 13,00672
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d32206d4bb9dbd12360d11d39b31237d54a8dcf8&vid=286a07a5e34a1ec55ff6339785af95d9a4326825&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR