Benzamidine hydrochloride hydrate, Sigma-Aldrich®

Leverandør: Merck
Warning

Synonymer: Phenylamidine hydrochloride hydrate, Benzene-1-carboximidamide hydrochloride hydrate, Benzenecarboximidamide hydrochloride hydrate, Benzamidinium chloride hydrate

M-Clarity™ quality level = MQ200

8.20122.0025 8.20122.1000 8.20122.0002
8.20122.0025EA 1180 NOK
8.20122.0025 8.20122.1000 8.20122.0002
Benzamidine hydrochloride hydrate, Sigma-Aldrich®
Benzamidine hydrochloride hydrate
Formel: C₆H₅C(=NH)NH₂·HCl·H₂O
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00066285
CAS-nummer: 206752-36-5
EINECS: 216-795-4

Bestill nå

Spesifikasjon Testresultater

Solubility (20 °C) soluble
Melting Point 80 - 84 °C
pH 5 (50 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR