Dimethyl succinate for synthesis, Sigma-Aldrich®

Leverandør: Merck
Warning

Synonymer: DBE-4 dibasic ester, Succinic acid dimethyl ester, 1,4-Butanedioic acid dimethyl ester

M-Clarity™ quality level = MQ200

8.20150.0250 8.20150.1000 8.20150.9020
8.20150.0250EA 421 NOK
8.20150.0250 8.20150.1000 8.20150.9020
Dimethyl succinate for synthesis, Sigma-Aldrich®
Dimethyl succinate
Formel: C₆H₁₀O₄
Kokepunkt: 200 °C (1013 hPa)
Smeltepunkt: 18…19 °C
Tetthet: 1,117 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00008466
CAS-nummer: 106-65-0
EINECS: 203-419-9
Merck Indeks: 12,09038

Bestill nå

Spesifikasjon Testresultater

Solubility 75 g/l (20 °C)
Melting Point 18 - 19 °C
Molar Mass 146.14 g/mol
Boiling Point 196 °C (1013 hPa)
Vapor Pressure 0.2 hPa (20 °C)
Flash Point 94 °C
Refractive Index 1.419 (20 °C, 589 nm)
Explosion Limit 1.0 - 8.5 %(V)
Density 1.12 g/cm3 (20 °C)
Ignition Point 365 °C

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR