tert-Butylbensen for synthesis, Sigma-Aldrich®

Leverandør: Merck
Danger


M-Clarity™ quality level = MQ200

8.20240.9025
8.20240.9025EA 0 NOK
8.20240.9025
tert-Butylbensen for synthesis, Sigma-Aldrich®
tert-Butylbensen
Formel: C₁₀H₁₄
MW: 134.22 g/mol
Kokepunkt: 169
Smeltepunkt: –58 °C
Tetthet: 0.8662 g/cm³ (20 °C)
Flash punkt: 44
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00008816
CAS-nummer: 98-06-6
EINECS: 202-632-4
FN: 2709
ADR: 3 III
Merck Indeks: 13,01550

Bestill nå

Spesifikasjon Testresultater

Solubility 29.5 mg/l (25 °C)
Melting Point -58 °C
Molar Mass 134.22 g/mol
Boiling Point 64 °C (27 hPa)
Vapor Pressure 1.33 hPa (13 °C)
Flash Point 44 °C
Refractive Index 1.492 (20 °C, 589 nm)
Explosion Limit 0.8 - 5.6 %(V)
Density 0.87 g/cm3 (20 °C)
Ignition Point 445 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR