Crotonoyl chloride cis- and trans-mixture for synthesis, Sigma-Aldrich®

Leverandør: Merck
Danger

Synonymer: But-2-enoyl chloride, 2-Butenoyl chloride, Crotonoyl Chloride, Crotonic acid chloride

M-Clarity™ quality level = MQ200

8.20351.0025
8.20351.0025EA 1240 NOK
8.20351.0025
Crotonoyl chloride cis- and trans-mixture for synthesis, Sigma-Aldrich®
Crotonoyl chloride
Formel: C₄H₅ClO
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00000734
CAS-nummer: 10487-71-5
EINECS: 234-010-3
FN: 2920
ADR: 8 (3),II

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR