Diethylamine hydrochloride for syntese, Sigma-Aldrich®

Leverandør: Merck
Danger


M-Clarity™ quality level = MQ200

8.20393.0005 8.20393.0250 8.20393.1000 8.20393.9025 8.20393.9050
8.20393.0005EA 354 NOK
8.20393.0005 8.20393.0250 8.20393.1000 8.20393.9025 8.20393.9050
Diethylamine hydrochloride for syntese, Sigma-Aldrich®
Diethylamine hydrochloride
Formel: C₄H₁₂ClN
Kokepunkt: 320 °C (1013 hPa)
Smeltepunkt: 220…225 °C
Tetthet: 1,0475 g/cm³ (21 °C)
MDL nummer: MFCD00012499
CAS-nummer: 660-68-4
EINECS: 211-541-9
FN: 3261
ADR: 8,II
Merck Indeks: 13,03138

Bestill nå

Spesifikasjon Testresultater

Solubility (20 °C) soluble
Melting Point 222 °C
Molar Mass 109.60 g/mol
Bulk Density 1100 kg/m3
Boiling Point 320 - 330 °C
pH 4.5 - 6.5 (10 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR