(2,4-Diklorofenoksy)eddiksyre, Sigma-Aldrich®

Leverandør: Merck
Danger

Synonymer: 2,4-D

M-Clarity™ quality level = MQ200

8.20451.0005 8.20451.0250 8.20451.1000 8.20451.9020
8.20451.0005EA 328 NOK
8.20451.0005 8.20451.0250 8.20451.1000 8.20451.9020
(2,4-Diklorofenoksy)eddiksyre, Sigma-Aldrich®
(2,4-Diklorofenoksy)eddiksyre
Formel: (C₆H₃Cl₂)OCH₂COOH
MW: 221.04 g/mol
Kokepunkt: 346
Smeltepunkt: 137…139 °C
Tetthet: 1.563 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00004300
CAS-nummer: 94-75-7
EINECS: 202-361-1
Merck Indeks: 13,02825

Bestill nå

Spesifikasjon Testresultater

Solubility 0.3 g/l (25 °C)
Melting Point 134 - 137 °C
Molar Mass 221.04 g/mol
Bulk Density 720 kg/m3
Boiling Point 160 °C (0.53 hPa)
Density 1.42 g/cm3 (25 °C)
Synonyms 2,4 D-acid
Hill Formula C8H6Cl2O3
Chemical Formula 2,4-(Cl)2C6H3OCH2COOH


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR