N,N'-Diphenyl-p-phenylenediamine for synthesis, Sigma-Aldrich®

Leverandør: Merck
Warning

Synonymer: DPPD, N,N′-Diphenyl-1,4-phenylenediamine, N,N'-Diphenyl-1,4-phenylenediamine

M-Clarity™ quality level = MQ200

8.20536.0250 8.20536.9010
8.20536.0250EA 1040 NOK
8.20536.0250 8.20536.9010
N,N'-Diphenyl-p-phenylenediamine for synthesis, Sigma-Aldrich®
N,N'-Diphenyl-p-phenylenediamine
Formel: C₁₈H₁₆N₂
Kokepunkt: 469 °C (1013 hPa)
Smeltepunkt: 146…149 °C
Tetthet: 1,21 g/cm³ (20 °C)
MDL nummer: MFCD00003015
CAS-nummer: 74-31-7
EINECS: 200-806-4
Merck Indeks: 12,03388

Bestill nå

Spesifikasjon Testresultater

Solubility (20 °C) practically insoluble
Melting Point 135 °C
Molar Mass 260.33 g/mol
Bulk Density 560 kg/m3
Boiling Point 220 - 225 °C (1.33 hPa)
Density 1.3 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR