DL-Malic acid disodium salt for synthesis, Sigma-Aldrich®

Leverandør: Merck

Synonymer: Di-Sodium Dl-Malate, DL-Malic acid disodium salt, DL-Apple acid disodium salt, Disodium DL-Malate hydrate

M-Clarity™ quality level = MQ200

8.20872.0025 8.20872.0100
8.20872.0025EA 766 NOK
8.20872.0025 8.20872.0100
DL-Malic acid disodium salt for synthesis, Sigma-Aldrich®
DL-Malic acid disodium salt
Formel: C₄H₄Na₂O₅
MDL nummer: MFCD00012466
CAS-nummer: 676-46-0
EINECS: 211-627-6

Bestill nå

Spesifikasjon Testresultater

Solubility (20 °C) soluble
Molar Mass 178.05 g/mol

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR