Diethyl-3-oxoglutarate

Leverandør: Merck

Synonymer: Diethyl 1,3-acetonedicarboxylate, Diethyl acetone-1,3-dicarboxylate

8.20941.0100
8.20941.0100EA 0 NOK
8.20941.0100
Diethyl-3-oxoglutarate
Diethyl-3-oxoglutarate
Formel: C₉H₁₄O₅
MW: 202.21 g/mol
MDL nummer: MFCD00009200
CAS-nummer: 105-50-0
EINECS: 203-302-2

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR