Trietylammoniumklorid, Sigma-Aldrich®

Leverandør: Merck
Danger

Synonymer: Triethylamine hydrochloride

M-Clarity™ quality level = MQ200

8.21135.0005 8.21135.0250 8.21135.1000 8.21135.9025
8.21135.0005EA 289 NOK
8.21135.0005 8.21135.0250 8.21135.1000 8.21135.9025
Trietylammoniumklorid, Sigma-Aldrich®
Trietylammoniumklorid
Formel: C₆H₁₆ClN
Tetthet: 0,7637 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00012500
CAS-nummer: 554-68-7
EINECS: 209-067-2
FN: 1759
ADR: 8,III

Bestill nå

Spesifikasjon Testresultater

Solubility 1440 g/l (20 °C)
Melting Point 255 - 258 °C
Molar Mass 137.65 g/mol
Bulk Density 220 kg/m3
Density 1.07 g/cm3 (21 °C)
pH 5 (10 g/l, H2O, 20 °C)
Synonyms Triethylamine hydrochloride
Hill Formula C6H16ClN
Chemical Formula (C2H5)3N * HCl


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR