Trimethyl phosphate zS, Sigma-Aldrich®

Leverandør: Merck
Danger

Synonymer: Trimethylphosphate

MQ200

8.21184.0100 8.21184.0500 8.21184.2500
8.21184.0100EA 355 NOK
8.21184.0100 8.21184.0500 8.21184.2500
Trimethyl phosphate zS, Sigma-Aldrich®
Trimethyl phosphate
Formel: C₃H₉O₄P
MW: 140.08 g/mol
Kokepunkt: 197.2
Smeltepunkt: –46 °C
Tetthet: 1.21 g/cm³ (20 °C)
Flash punkt: >150 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00008348
CAS-nummer: 512-56-1
EINECS: 208-144-8

Bestill nå

Spesifikasjon Testresultater

Solubility 1000 g/l (25 °C)
Melting Point -46 °C
Molar Mass 140.08 g/mol
Boiling Point 197 °C (1013 hPa)
Vapor Pressure 1.133 hPa (25 °C)
Flash Point 150 °C
Refractive Index 1.3967 (20 °C)
Density 1.21 g/cm3 (20 °C)

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR