1-Adamantanecarbonitrile

Leverandør: Merck

Synonymer: 1-Cyanotricyclo[3.3.1.1(3,7)]decane, Tricyclo[3.3.1.1(3,7)]decane-1-carbonitrile, 1-Cyanoadamantane

8.21657.0005
8.21657.0005EA 0 NOK
8.21657.0005
1-Adamantanecarbonitrile
1-Adamantanecarbonitrile
Formel: C₁₁H₁₅N
MW: 161.25 g/mol
Kokepunkt: 275
Smeltepunkt: 185…187 °C
Tetthet: 1.13 g/cm³ (25 °C)
MDL nummer: MFCD00074731
CAS-nummer: 23074-42-2
EINECS: 245-413-9
FN: 3439
ADR: 6.1 III

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR