N,N-Dimetyl-p-toluidine for syntese, Sigma-Aldrich®

Leverandør: Merck
Danger


M-Clarity™ quality level = MQ200

8.22040.0005 8.22040.0250 8.22040.1000 8.22040.9025
8.22040.0005EA 331 NOK
8.22040.0005 8.22040.0250 8.22040.1000 8.22040.9025
N,N-Dimetyl-p-toluidine for syntese, Sigma-Aldrich®
N,N-Dimetyl-p-toluidine
Formel: C₉H₁₃N
Kokepunkt: 211 °C (1013 hPa)
Smeltepunkt: –15 °C
Tetthet: 0,9 g/cm³ (20 °C)
Flash punkt: 76 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00008316
CAS-nummer: 99-97-8
EINECS: 202-805-4
FN: 2810
ADR: 6.1,III

Bestill nå

Spesifikasjon Testresultater

Solubility 0.65 g/l (37 °C)
Melting Point -15 °C
Molar Mass 135.21 g/mol
Boiling Point 215 °C (1013 hPa)
Vapor Pressure 0.1 hPa (20 °C)
Flash Point 76 °C
Refractive Index 1.546 (20 °C, 589 nm)
Density 0.94 g/cm3 (20 °C)
Ignition Point 425 °C
Synonyms 4-Dimethylaminotoluene
Hill Formula C9H13N
Chemical Formula 4-(CH3)C6H4N(CH3)2

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR