Morpholine hydrochloride

Leverandør: Merck

Synonymer: Morpholinium chloride

MQ200

8.22101.0250
8.22101.0250EA 1800 NOK
8.22101.0250
Morpholine hydrochloride
Morpholine hydrochloride
Formel: C₄H₁₀ClNO
MW: 123.58 g/mol
MDL nummer: MFCD00035287
CAS-nummer: 10024-89-2
EINECS: 233-029-4

Bestill nå

Spesifikasjon Testresultater

Solubility (20 °C) soluble
Melting Point 174 - 178 °C
Molar Mass 123.58 g/mol


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR