Sodium sulphanilate dihydrate sodium salt for synthesis, Sigma-Aldrich®

Leverandør: Merck

Synonymer: 4-Aminobenzenesulphonic acid, sodium salt dihydrate

M-Clarity™ quality level = MQ200

8.22145.0250
8.22145.0250EA 1060 NOK
8.22145.0250
Sodium sulphanilate dihydrate sodium salt for synthesis, Sigma-Aldrich®
Sodium sulfanilate dihydrate
Formel: C₆H₁₀NNaO₅S
MW: 231.198 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00064394
CAS-nummer: 6106-22-5
EINECS: 208-208-5

Bestill nå

Spesifikasjon Testresultater

Solubility 170 g/l (20 °C)
Molar Mass 231.2 g/mol
Chemical formula 4-(NH2)C6H4SO3Na*2H2O
Molecular weight 231,2 g/mol
Synonyms Sulfanilic acid sodium salt
Identity (IR) passes test
Storage temperature +15...+25 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR