Methyl D-glucopyranoside (mixture of anomers) for synthesis, Sigma-Aldrich®

Leverandør: Merck


M-Clarity™ quality level = MQ200

8.40024.0005 8.40024.0100
8.40024.0005EA 330 NOK
8.40024.0005 8.40024.0100
Methyl D-glucopyranoside (mixture of anomers) for synthesis, Sigma-Aldrich®
Methyl D-glucopyranoside (mixture of anomers)
Formel: C₇H₁₄O₆
CAS-nummer: 3149-68-6
EINECS: 221-581-9

Bestill nå

Spesifikasjon Testresultater

Solubility 600 g/l (25 °C)
Melting Point 155 - 165 °C
Molar Mass 194.18 g/mol
Bulk Density 400 kg/m3

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR