N,O-Dimethylhydroxylamine hydrochloride for synthesis, Sigma-Aldrich®

Leverandør: Merck
Warning


M-Clarity™ quality level = MQ200

8.40074.0010 8.40074.0050
8.40074.0010EA 753 NOK
8.40074.0010 8.40074.0050
N,O-Dimethylhydroxylamine hydrochloride for synthesis, Sigma-Aldrich®
N,O-Dimethylhydroxylamine hydrochloride
Formel: C₂H₈ClNO
MW: 97.55 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00012485
CAS-nummer: 6638-79-5
EINECS: 229-642-1

Bestill nå

Spesifikasjon Testresultater

Solubility (20 °C) soluble
Melting Point 112 - 116 °C
Molar Mass 97.54 g/mol


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR