Acetonyltriphenylphosphonium chloride for synthesis, Sigma-Aldrich®

Leverandør: Merck

Synonymer: (2-Oxopropyl)triphenylphosphonium chloride

M-Clarity™ quality level = MQ200

8.41028.0025
8.41028.0025EA 1125 NOK
8.41028.0025
Acetonyltriphenylphosphonium chloride for synthesis, Sigma-Aldrich®
Acetonyltriphenylphosphonium chloride
Formel: C₂₁H₂₀ClOP
MW: 354.82 g/mol
Smeltepunkt: 243…245 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011813
CAS-nummer: 1235-21-8
EINECS: 214-974-1

Bestill nå

Spesifikasjon Testresultater

Melting Point 243 - 245 °C
Molar Mass 354.81 g/mol
Synonyms 2-Oxopropyl triphenylphosphonium chloride
Hill Formula C21H20ClOP


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR