Acetaldehyde oxime (mixture of syn and anti) for synthesis, Sigma-Aldrich®

Leverandør: Merck
Danger

Synonymer: Acetaldoxime

M-Clarity™ quality level = MQ200

8.41085.0050 8.41085.0250
8.41085.0050EA 876 NOK
8.41085.0050 8.41085.0250
Acetaldehyde oxime (mixture of syn and anti) for synthesis, Sigma-Aldrich®
Acetaldehyde oxime (mixture of syn and anti)
Formel: C₂H₅NO
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00002124
CAS-nummer: 107-29-9
EINECS: 203-479-6
FN: 2332
ADR: 3,III

Bestill nå

Spesifikasjon Testresultater

Solubility 185 g/l (25 °C)
Molar Mass 59.07 g/mol
Boiling Point 115 °C
Vapor Pressure 13 hPa (25 °C)
Flash Point 40 °C
Explosion Limit 4.2 - 50 %(V)
Density 0.969 g/cm3 (20 °C)
Ignition Point 380 °C

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR