Methylenediphosphonic acid, Sigma-Aldrich®

Leverandør: Merck
Danger

Synonymer: Medronic acid

M-Clarity™ quality level = MQ200

8.41124.0001
8.41124.0001EA 1190 NOK
8.41124.0001
Methylenediphosphonic acid, Sigma-Aldrich®
Methylenediphosphonic acid
Formel: CH₆O₆P₂
MW: 176 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00002139
CAS-nummer: 1984-15-2
EINECS: 217-851-0
FN: 3261
ADR: 8 II

Bestill nå

Spesifikasjon Testresultater

Melting Point 200 - 205 °C
Molar Mass 176 g/mol


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR