Triisopropylsilane for synthesis, Sigma-Aldrich®

Leverandør: Merck
Warning


M-Clarity™ quality level = MQ200

8.41359.0010
8.41359.0010EA 536 NOK
8.41359.0010
Triisopropylsilane for synthesis, Sigma-Aldrich®
Triisopropylsilane
Formel: C₉H₂₂Si
Kokepunkt: 84 °C (1013 hPa)
Tetthet: 0,7734 g/cm³ (20 °C)
Flash punkt: 35 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00009657
CAS-nummer: 6485-79-6
FN: 1993
ADR: 3,II

Bestill nå

Spesifikasjon Testresultater

Molar Mass 158.35 g/mol
Boiling Point 84 - 85 °C (46.7 hPa)
Flash Point 35 °C
Density 0.77 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR