4-Piperidone monohydrate hydrochloride for synthesis, Sigma-Aldrich®

Leverandør: Merck

Synonymer: 4,4-Piperidinediol Hydrochloride, 4,4-Dihydroxypiperidine Hydrochloride, 4-Ketopiperidine Monohydrate Hydrochloride, 4-Piperidinone monohydrate hydrochloride, Piperidin-4-one hydrate hydrochloride, 4-Azacyclohexanone Monohydrate Hydrochloride

M-Clarity™ quality level = MQ200

8.41672.0010 8.41672.0050
8.41672.0010EA 492 NOK
8.41672.0010 8.41672.0050
4-Piperidone monohydrate hydrochloride for synthesis, Sigma-Aldrich®
4-Piperidone monohydrate hydrochloride
Formel: C₅H₉NO·xHCl·yH₂O
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00012776
CAS-nummer: 320589-77-3
EINECS: 255-602-8

Bestill nå

Spesifikasjon Testresultater

Melting Point 95...99 °C
Molar Mass 153,61 g/mol
Hill Formula C5H9NO*ClH*H2O
HS Code 2933 39 99
Storage temperature 15...25 °C
Assay (argentometric) ≥98% (m)
Melting range (lower value) ≥91 °C
Melting range (upper value) ≤94 °C
Identity (IR) conforms


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR