DL-Carnitine hydrochloride for synthesis, Sigma-Aldrich®

Leverandør: Merck

Synonymer: 3-Hydroxy-4-(trimethylammonio)butanoate hydrochloride

M-Clarity™ quality level = MQ200

8.41774.0025
8.41774.0025EA 601 NOK
8.41774.0025
DL-Carnitine hydrochloride for synthesis, Sigma-Aldrich®
DL-Carnitine hydrochloride
Formel: C₇H₁₆ClNO₃
MDL nummer: MFCD00011904
CAS-nummer: 461-05-2
EINECS: 207-309-1

Bestill nå

Spesifikasjon Testresultater

Solubility (20 °C) soluble
Melting Point 196 - 197 °C decomposes
Molar Mass 197.66 g/mol
pH 1.8 - 2.5 (100 g/l, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR