Sodium-1-hexadecanesulphonate for synthesis, Sigma-Aldrich®

Leverandør: Merck

Synonymer: Sodium hexadecane-1-sulphonate, Hexadecane-1-sulphonic acid sodium salt

M-Clarity™ quality level = MQ200

8.41813.0010
8.41813.0010EA 1975 NOK
8.41813.0010
Sodium-1-hexadecanesulphonate for synthesis, Sigma-Aldrich®
Sodium-1-hexadecanesulphonate
Formel: C₁₆H₃₃NaSO₃
MW: 328.49 g/mol
MDL nummer: MFCD00007528
CAS-nummer: 15015-81-3
EINECS: 239-100-6

Bestill nå

Spesifikasjon Testresultater

Melting Point 265.3 °C
Molar Mass 328.48 g/mol
Hill Formula C16H33NaO3S


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR