1,3-Dimethylbarbituric acid for syntese, Sigma-Aldrich®

Leverandør: Merck
Danger


M-Clarity™ quality level = MQ200

8.42116.0005 8.42116.0100
8.42116.0005EA 518 NOK
8.42116.0005 8.42116.0100
1,3-Dimethylbarbituric acid for syntese, Sigma-Aldrich®
1,3-Dimethylbarbituric acid
Formel: C₆H₈N₂O₃
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00006675
CAS-nummer: 769-42-6
EINECS: 212-211-7

Bestill nå

Spesifikasjon Testresultater

Solubility 60 g/l (20 °C)
Melting Point 120 - 124 °C
Molar Mass 156.14 g/mol
Bulk Density 500 kg/m3
pH 2.5 (100 g/l, H2O, 20 °C) (slurry)

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR