Cyanoacetylurea ≥98.0% (by HPLC) for synthesis

Leverandør: Merck
Danger

Synonymer: N-Carbamoyl-2-cyanoacetamide, Acetamide, N-(aminocarbonyl)-2-cyano-

M-Clarity™ quality level = MQ200

8.43302.0025
8.43302.0025EA 485 NOK
8.43302.0025
Cyanoacetylurea ≥98.0% (by HPLC) for synthesis
Cyanoacetylurea
Formel: C₄H₅N₃O₂
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00007947
CAS-nummer: 1448-98-2
EINECS: 215-906-3
FN: 3439
ADR: 6.1,III

Bestill nå

Spesifikasjon Testresultater

Assay (HPLC, area%) ≥ 98% (a/a)
Melting range (lower value) ≥ 203 °C
Melting range (upper value) ≤ 207 °C
Identity (IR) passes test
HS Code 2926 90 95
Melting Point 203...207 °C
Storage 15...25 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR